2-Amino-6-bromoquinazolin-4-ol
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
OR300184-1G
OR300184-250MG
APOSOR300184-1GEA
343
EUR
APOSOR300184-1G
APOSOR300184-250MG
2-Amino-6-bromoquinazolin-4-ol
2-Amino-6-bromoquinazolin-4-ol
Formule:
C₈H₆BrN₃O Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD16250251 CAS nummer: 130148-53-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...