1,2,5,6-Tetrabromocyclooctane

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

OR302173-1G OR302173-250MG
APOSOR302173-1GEA 1460 EUR
APOSOR302173-1G APOSOR302173-250MG
1,2,5,6-Tetrabromocyclooctane
1,2,5,6-Tetrabromocyclooctane
Formule: C₈H₁₂Br₄
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00666115
CAS nummer: 3194-57-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR