5-Chloro-2-methoxyphenylhydrazine hydrochloride
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
OR3385-1G
OR3385-250MG
APOSOR3385-1GEA
125
EUR
APOSOR3385-1G
APOSOR3385-250MG
5-Chloro-2-methoxyphenylhydrazine hydrochloride
5-Chloro-2-methoxyphenylhydrazine hydrochloride
Formule:
C₇H₉ClN₂O·ClH Smeltpunt: 192…196 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD03196500 CAS nummer: 5446-16-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...