2-Propylidenemalononitrile 95+%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OR346446-1G OR346446-5G
APOSOR346446-1GEA 245 EUR
APOSOR346446-1G APOSOR346446-5G
2-Propylidenemalononitrile 95+%
2-Propylidenemalononitrile
Formule: C₆H₆N₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD06213931
CAS nummer: 52833-34-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR