(9H-Xanthen-9-yl)methanol ≥98%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OR370044-100MG
OR370044-500MG
APOSOR370044-100MGEA
406
EUR
APOSOR370044-100MG
APOSOR370044-500MG
(9H-Xanthen-9-yl)methanol ≥98%
(9H-Xanthen-9-yl)methanol
Formule:
C₁₄H₁₂O₂ Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00100363 CAS nummer: 5490-92-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...