5-(Bromomethyl)isoquinoline hydrobromide

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

OR42118-1G OR42118-250MG
APOSOR42118-1GEA 0 EUR
APOSOR42118-1G APOSOR42118-250MG
5-(Bromomethyl)isoquinoline hydrobromide
5-(Bromomethyl)isoquinoline hydrobromide
Formule: C₁₀H₈BrN·BrH
MW: 302.993 g/mol
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD11976354
CAS nummer: 586373-76-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR