b-Amino-cyclohexaneethanol hydrochloride 97%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OR452144-1G
OR452144-250MG
OR452144-500MG
APOSOR452144-1GEA
723
EUR
APOSOR452144-1G
APOSOR452144-250MG
APOSOR452144-500MG
b-Amino-cyclohexaneethanol hydrochloride 97%
b-Amino-cyclohexaneethanol hydrochloride
Formule:
C₈H₁₇NO·ClH Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD20731221 CAS nummer: 99839-72-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...