2-Bromo-5-iodoaniline, N-BOC protected
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 2-Bromo-N-(tert-butoxycarbonyl)-5-iodoaniline, tert-Butyl (2-bromo-5-iodophenyl)carbamate
OR46082-1G OR46082-250MG OR46082-5G
APOSOR46082-1GEA 211 EUR
APOSOR46082-1G APOSOR46082-250MG APOSOR46082-5G
2-Bromo-5-iodoaniline, N-BOC protected
2-Bromo-5-iodoaniline, N-BOC protected
Formule: C₁₁H₁₃BrINO₂
MW: 398.035 g/mol
Smeltpunt: 127…132 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD23704427
CAS nummer: 221876-99-9