7-Bromo-1,4-dihydro-3,1-benzoxazin-2-one

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OR471595-1G OR471595-250MG OR471595-5G
APOSOR471595-1GEA 400 EUR
APOSOR471595-1G APOSOR471595-250MG APOSOR471595-5G
7-Bromo-1,4-dihydro-3,1-benzoxazin-2-one
7-Bromo-1,4-dihydro-3,1-benzoxazin-2-one
Formule: C₈H₆BrNO₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD12755795
CAS nummer: 1245808-46-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR