(S)-1-Isopropyl-2-methylpiperazine 95%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OR471725-1G
OR471725-250MG
APOSOR471725-1GEA
1630
EUR
APOSOR471725-1G
APOSOR471725-250MG
(S)-1-Isopropyl-2-methylpiperazine 95%
(S)-1-Isopropyl-2-methylpiperazine
Formule:
C₈H₁₈N₂ Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD09264178 CAS nummer: 807319-94-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...