3-Bromo-4-cyclopropylnitrobenzene
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
OR47844-1G
OR47844-250MG
OR47844-5G
APOSOR47844-1GEA
526
EUR
APOSOR47844-1G
APOSOR47844-250MG
APOSOR47844-5G
3-Bromo-4-cyclopropylnitrobenzene
3-Bromo-4-cyclopropylnitrobenzene
Formule:
C₉H₈BrNO₂ MW: 242.069 g/mol Bewaar temperatuur: Vriezer |
MDL Nummer:
MFCD28399181 CAS nummer: 57807-35-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...