N-Boc-6-Bromohexylamine
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
tert-Butyl (6-bromohexyl)carbamate, Boc-NH-C6-Br
OR55147-1G
OR55147-5G
APOSOR55147-1GEA
29.2
EUR
APOSOR55147-1G
APOSOR55147-5G
N-Boc-6-Bromohexylamine
N-Boc-6-Bromohexylamine
Building block for preparing analogs of biologically active compounds
Formule:
C₁₁H₂₂BrNO₂ Dichtheid: 1,192 g/cm³ (25 °C) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD06201019 CAS nummer: 142356-33-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...