5-(Bromomethyl)-2-iodophenol 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

OR55567-1G OR55567-250MG OR55567-5G
APOSOR55567-1GEA 497 EUR
APOSOR55567-1G APOSOR55567-250MG APOSOR55567-5G
5-(Bromomethyl)-2-iodophenol 95%
5-(Bromomethyl)-2-iodophenol
Formule: C₇H₆BrIO
MW: 312.93 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD25459292
CAS nummer: 1823881-56-6
UN: 3261
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR