2,4,5-Trichlorobenzyl bromide

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

OR61083-1G OR61083-250MG
APOSOR61083-1GEA 117 EUR
APOSOR61083-1G APOSOR61083-250MG
2,4,5-Trichlorobenzyl bromide
2,4,5-Trichlorobenzyl bromide
Formule: C₇H₄BrCl₃
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD09999821
CAS nummer: 81778-11-2
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR