3-Amino-5-(hydrazonomethyl)-1H-indazole 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OR61395-1G OR61395-500MG
APOSOR61395-1GEA 338 EUR
APOSOR61395-1G APOSOR61395-500MG
3-Amino-5-(hydrazonomethyl)-1H-indazole 97%
3-Amino-5-(hydrazonomethyl)-1H-indazole
Formule: C₈H₉N₅
Smeltpunt: >300 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD19381756
CAS nummer: 1312142-38-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR