N-Butyl-4-hydroxybenzamide 98%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OR900653-1G
OR900653-5G
APOSOR900653-1GEA
184
EUR
APOSOR900653-1G
APOSOR900653-5G
N-Butyl-4-hydroxybenzamide 98%
N-Butyl-4-hydroxybenzamide
Formule:
C₁₁H₁₅NO₂ Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD01118983 CAS nummer: 60696-49-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...