2-Cyclobutyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OR901542-1G
OR901542-250MG
OR901542-5G
APOSOR901542-1GEA
178
EUR
APOSOR901542-1G
APOSOR901542-250MG
APOSOR901542-5G
2-Cyclobutyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97%
2-Cyclobutyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formule:
C₁₀H₁₉BO₂ Bewaar temperatuur: Vriezer |
MDL Nummer:
MFCD22392290 CAS nummer: 1360914-08-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...