2-(Ethylsulphanyl)aniline 95%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OR902320-10G
OR902320-1G
OR902320-5G
APOSOR902320-10GEA
894
EUR
APOSOR902320-10G
APOSOR902320-1G
APOSOR902320-5G
2-(Ethylsulphanyl)aniline 95%
2-(Ethylsulphanyl)aniline
Formule:
C₈H₁₁NS MW: 153.245 g/mol Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD08691983 CAS nummer: 13920-91-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...