N-Ethyl-3-hydroxybenzamide 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OR908144-10G OR908144-1G OR908144-5G
APOSOR908144-10GEA 760 EUR
APOSOR908144-10G APOSOR908144-1G APOSOR908144-5G
N-Ethyl-3-hydroxybenzamide 98%
N-Ethyl-3-hydroxybenzamide
Formule: C₉H₁₁NO₂
MW: 165.189 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD08729290
CAS nummer: 15788-98-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR