8-Nitroquinolin-2(1H)-one 95%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OR925748-1G
OR925748-250MG
APOSOR925748-1GEA
465
EUR
APOSOR925748-1G
APOSOR925748-250MG
8-Nitroquinolin-2(1H)-one 95%
8-Nitroquinolin-2(1H)-one
Formule:
C₉H₆N₂O₃ MW: 190.156 g/mol Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD21605405 CAS nummer: 7461-12-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...