3'-(N-Methylacetyl)acetophenone 95%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OR926748-100MG
OR926748-1G
OR926748-250MG
APOSOR926748-100MGEA
155
EUR
APOSOR926748-100MG
APOSOR926748-1G
APOSOR926748-250MG
3'-(N-Methylacetyl)acetophenone 95%
3'-(N-Methylacetyl)acetophenone
Formule:
C₉H₁₁NO Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD16470104 CAS nummer: 42865-75-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...