4-Methyl-5-pyrimidinecarbonitrile 95%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OR928387-100MG
OR928387-1G
OR928387-250MG
APOSOR928387-100MGEA
162
EUR
APOSOR928387-100MG
APOSOR928387-1G
APOSOR928387-250MG
4-Methyl-5-pyrimidinecarbonitrile 95%
4-Methyl-5-pyrimidinecarbonitrile
Formule:
C₆H₅N₃ Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD19982748 CAS nummer: 1150633-16-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...