3,5-Dimethoxypyridine 98%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OR932195-1G
OR932195-5G
APOSOR932195-1GEA
0
EUR
APOSOR932195-1G
APOSOR932195-5G
3,5-Dimethoxypyridine 98%
3,5-Dimethoxypyridine
Formule:
C₇H₉NO₂ Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD01646188 CAS nummer: 18677-48-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...