1-Ethylpiperidine-3-carbaldehyde hydrochloride 95%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OR932717-1G
OR932717-250MG
APOSOR932717-1GEA
541
EUR
APOSOR932717-1G
APOSOR932717-250MG
1-Ethylpiperidine-3-carbaldehyde hydrochloride 95%
1-Ethylpiperidine-3-carbaldehyde hydrochloride
Formule:
C₈H₁₅NO·ClH Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD09864359 CAS nummer: 1255717-79-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...