Ethyl 2-(cyclopropylamino)acetate 95%
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OR940640-1G OR940640-250MG
APOSOR940640-1GEA 380 EUR
APOSOR940640-1G APOSOR940640-250MG
Ethyl 2-(cyclopropylamino)acetate 95%
Ethyl 2-(cyclopropylamino)acetate
Formule: C₇H₁₃NO₂
MDL Nummer: MFCD11149655
CAS nummer: 71922-62-8