3-Cyano-N-isopropylbenzamide 96%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OR946460-1G OR946460-5G
APOSOR946460-1GEA 220 EUR
APOSOR946460-1G APOSOR946460-5G
3-Cyano-N-isopropylbenzamide 96%
3-Cyano-N-isopropylbenzamide
Formule: C₁₁H₁₂N₂O
MW: 188.226 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD09940478
CAS nummer: 623569-57-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR