3-Methanesulphonyl-phenol 97%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OR951080-1G
OR951080-250MG
APOSOR951080-1GEA
584
EUR
APOSOR951080-1G
APOSOR951080-250MG
3-Methanesulphonyl-phenol 97%
3-Methanesulphonyl-phenol
Formule:
C₇H₈O₃S Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD11839805 CAS nummer: 14763-61-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...