3-tert-Butyl-4-hydroxyfuran-2(5H)-one 95%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OR951398-1G
OR951398-250MG
APOSOR951398-1GEA
811
EUR
APOSOR951398-1G
APOSOR951398-250MG
3-tert-Butyl-4-hydroxyfuran-2(5H)-one 95%
3-tert-Butyl-4-hydroxyfuran-2(5H)-one
Formule:
C₈H₁₂O₃ Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD08236783 CAS nummer: 114908-93-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...