5-Ethyl-2-pyridine alcohol 95%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OR951862-1G
OR951862-250MG
APOSOR951862-1GEA
892
EUR
APOSOR951862-1G
APOSOR951862-250MG
5-Ethyl-2-pyridine alcohol 95%
5-Ethyl-2-pyridine alcohol
Formule:
C₇H₉NO Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD06658539 CAS nummer: 53428-03-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...