N-Isopropylbenzenesulphonamide 95%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OR958001-1G
OR958001-250MG
APOSOR958001-1GEA
166
EUR
APOSOR958001-1G
APOSOR958001-250MG
N-Isopropylbenzenesulphonamide 95%
N-Isopropylbenzenesulphonamide
Formule:
C₉H₁₃NO₂S Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00591077 CAS nummer: 5339-69-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...