2-Methyl-7-nitro-3,4-dihydro-2H-1,4-benzoxazine 98%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OR960207-1G
OR960207-5G
APOSOR960207-1GEA
220
EUR
APOSOR960207-1G
APOSOR960207-5G
2-Methyl-7-nitro-3,4-dihydro-2H-1,4-benzoxazine 98%
2-Methyl-7-nitro-3,4-dihydro-2H-1,4-benzoxazine
Formule:
C₉H₁₀N₂O₃ Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD23379851 CAS nummer: 941291-25-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...