5-Methanesulphonylpyridine-2-carbonitrile 98%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OR960346-1G
OR960346-250MG
APOSOR960346-1GEA
636
EUR
APOSOR960346-1G
APOSOR960346-250MG
5-Methanesulphonylpyridine-2-carbonitrile 98%
5-Methanesulphonylpyridine-2-carbonitrile
Formule:
C₇H₆N₂O₂S Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD26395889 CAS nummer: 848141-13-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...