1-Cyclopropyl-1-(4-pyridyl)methylamine 95%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OR965312-1G
OR965312-250MG
APOSOR965312-1GEA
490
EUR
APOSOR965312-1G
APOSOR965312-250MG
1-Cyclopropyl-1-(4-pyridyl)methylamine 95%
1-Cyclopropyl-1-(4-pyridyl)methylamine
Formule:
C₉H₁₂N₂ Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD18652126 CAS nummer: 1270506-12-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...