Ethyl 1H-tetrafluoropropanaote 98%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PC10059-25G
PC10059-100G
APOSPC10059-25GEA
148
EUR
APOSPC10059-25G
APOSPC10059-100G
Ethyl 1H-tetrafluoropropanaote 98%
Ethyl 1H-tetrafluoropropanaote
Formule:
C₅H₆F₄O₂ |
MDL Nummer:
MFCD04039197 CAS nummer: 399-92-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...