2-Bromo-6-fluorobenzenesulphonamide
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
PC10330-1G
PC10330-250MG
PC10330-5G
APOSPC10330-1GEA
491
EUR
APOSPC10330-1G
APOSPC10330-250MG
APOSPC10330-5G
2-Bromo-6-fluorobenzenesulphonamide
2-Bromo-6-fluorobenzenesulphonamide
Formule:
C₆H₅BrFNO₂S Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD07368674 CAS nummer: 886762-60-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...