Methyl 2-fluoropropanoate 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

PC19689-1G PC19689-25G PC19689-5G
APOSPC19689-1GEA 95 EUR
APOSPC19689-1G APOSPC19689-25G APOSPC19689-5G
Methyl 2-fluoropropanoate 98%
Methyl 2-fluoropropanoate
Formule: C₄H₇FO₂
Kookpunt: 107…108 °C
Dichtheid: 1,0806 g/cm³ (25 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD04972939
CAS nummer: 2366-56-5
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR