Silver(I) pentafluoropropanoate
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
PC1983-5G
PC1983-1G
APOSPC1983-5GEA
144
EUR
APOSPC1983-5G
APOSPC1983-1G
Silver(I) pentafluoropropanoate
Silver(I) pentafluoropropanoate
Formule:
C₃F₅O₂·Ag Smeltpunt: 242…244 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00013251 CAS nummer: 509-09-1 EINECS: 208-093-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...