2-Methoxy-4-(trifluoromethyl)pyridine 98%
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PC201198-1G PC201198-5G
APOSPC201198-1GEA 205 EUR
APOSPC201198-1G APOSPC201198-5G
2-Methoxy-4-(trifluoromethyl)pyridine 98%
2-Methoxy-4-(trifluoromethyl)pyridine
Formule: C₇H₆F₃NO
MDL Nummer: MFCD00096939
CAS nummer: 219715-34-1