5-(3-Fluorophenyl)-2-methylpyridine 95+%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PC201289-1G
PC201289-25G
PC201289-5G
APOSPC201289-1GEA
0
EUR
APOSPC201289-1G
APOSPC201289-25G
APOSPC201289-5G
5-(3-Fluorophenyl)-2-methylpyridine 95+%
5-(3-Fluorophenyl)-2-methylpyridine
Formule:
C₁₂H₁₀FN Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD13152238 CAS nummer: 713143-67-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...