3-Ethyl-4-fluorobenzonitrile 95+%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PC201333-1G
PC201333-25G
PC201333-5G
APOSPC201333-1GEA
122
EUR
APOSPC201333-1G
APOSPC201333-25G
APOSPC201333-5G
3-Ethyl-4-fluorobenzonitrile 95+%
3-Ethyl-4-fluorobenzonitrile
Formule:
C₉H₈FN Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD19441199 CAS nummer: 869299-63-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...