3-Ethynyl-2-fluoropyridine 95%
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PC201379-100MG PC201379-250MG PC201379-1G
APOSPC201379-100MGEA 92.1 EUR
APOSPC201379-100MG APOSPC201379-250MG APOSPC201379-1G
3-Ethynyl-2-fluoropyridine 95%
3-Ethynyl-2-fluoropyridine
Formule: C₇H₄FN
MDL Nummer: MFCD13175283
CAS nummer: 933768-06-0