7-Amino-5-fluoroindolin-2-one 95+%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PC201382-100MG PC201382-250MG
APOSPC201382-100MGEA 26 EUR
APOSPC201382-100MG APOSPC201382-250MG
7-Amino-5-fluoroindolin-2-one 95+%
7-Amino-5-fluoroindolin-2-one
Formule: C₈H₇FN₂O
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD13193359
CAS nummer: 945381-62-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR