1-[(4E)-5-(Dimethylamino)-3-oxopent-4-en-2-yl]-5-(trifluoromethyl)-1,2-dihydropyridin-2-one

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PC210073-1G PC210073-500MG
APOSPC210073-1GEA 900 EUR
APOSPC210073-1G APOSPC210073-500MG
1-[(4E)-5-(Dimethylamino)-3-oxopent-4-en-2-yl]-5-(trifluoromethyl)-1,2-dihydropyridin-2-one
1-[(4E)-5-(Dimethylamino)-3-oxopent-4-en-2-yl]-5-(trifluoromethyl)-1,2-dihydropyridin-2-one
Formule: C₁₃H₁₅F₃N₂O₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD28347972
CAS nummer: 1644602-78-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR