1-Bromo-3-(trifluoromethyl)-1,1,4,4,4-pentafluorobut-2-ene 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PC2548-1G PC2548-5G
APOSPC2548-1GEA 177 EUR
APOSPC2548-1G APOSPC2548-5G
1-Bromo-3-(trifluoromethyl)-1,1,4,4,4-pentafluorobut-2-ene 97%
1-Bromo-3-(trifluoromethyl)-1,1,4,4,4-pentafluorobut-2-ene
Formule: C₅HBrF₈
MW: 292.953 g/mol
Kookpunt: 74…76 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD07784212
CAS nummer: 885276-66-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR