3,8-Dichloro-6-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridine
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PC28300-1G PC28300-250MG
APOSPC28300-1GEA 222 EUR
APOSPC28300-1G APOSPC28300-250MG
3,8-Dichloro-6-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridine
3,8-Dichloro-6-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridine
Formule: C₇H₂Cl₂F₃N₃
MDL Nummer: MFCD16876986
CAS nummer: 1823182-47-3