2',3'-Difluoropropiophenone 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 1-(2,3-Difluorophenyl)propan-1-one

PC2879-10G PC2879-1G PC2879-5G
APOSPC2879-10GEA 129 EUR
APOSPC2879-10G APOSPC2879-1G APOSPC2879-5G
2',3'-Difluoropropiophenone 97%
2',3'-Difluoropropiophenone
Formule: C₉H₈F₂O
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00061141
CAS nummer: 236754-63-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR