N,2-Dimethyl-6-(trifluoromethyl)nicotinamide

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PC300669-1G PC300669-5G
APOSPC300669-1GEA 234 EUR
APOSPC300669-1G APOSPC300669-5G
N,2-Dimethyl-6-(trifluoromethyl)nicotinamide
N,2-Dimethyl-6-(trifluoromethyl)nicotinamide
Formule: C₉H₉F₃N₂O
MW: 218.176 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD11826496
CAS nummer: 1090043-67-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR