4-Fluoro-3,5-dimethylphenylacetonitrile 96%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
PC302216-1G
PC302216-5G
APOSPC302216-1GEA
80.4
EUR
APOSPC302216-1G
APOSPC302216-5G
4-Fluoro-3,5-dimethylphenylacetonitrile 96%
4-Fluoro-3,5-dimethylphenylacetonitrile
Formule:
C₁₀H₁₀FN Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD09832374 CAS nummer: 1000513-61-0 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...