4-Bromo-2-fluoro-1-cyclopropylbenzene

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

PC32950-1G PC32950-250MG
APOSPC32950-1GEA 547 EUR
APOSPC32950-1G APOSPC32950-250MG
4-Bromo-2-fluoro-1-cyclopropylbenzene
4-Bromo-2-fluoro-1-cyclopropylbenzene
Formule: C₉H₈BrF
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD21099968
CAS nummer: 1353855-10-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR