3-Chloro-2-fluorobenzoyl fluoride 98%
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
PC4272-1G
PC4272-250MG
APOSPC4272-1GEA
243
EUR
APOSPC4272-1G
APOSPC4272-250MG
3-Chloro-2-fluorobenzoyl fluoride 98%
3-Chloro-2-fluorobenzoyl fluoride
Formule:
C₇H₃ClF₂O Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD04972721 CAS nummer: 85345-74-0 UN: 1759 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...